This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01595384_m1
See other ZC3H7B GE Assays ›
Gene Symbol:
ZC3H7B
Gene Name:
zinc finger CCCH-type containing 7B
Gene Aliases:
RoXaN
Chromosome Location:
Chr.22: 41301503 - 41360147 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01595384_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZC3H7B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017590.5 NP_060060.3 14 - 15 1944 128
XM_011530016.2 XP_011528318.1 14 - 15 1866 128
GenBank mRNA AB028954.1 - 11 - 12 1557 128
AF188530.1 - 14 - 15 1772 128
AK315009.1 - 14 - 15 1886 128
BC024313.2 - 5 - 6 611 128
BC098378.1 - 14 - 15 1882 128
BC152558.1 - 14 - 15 1922 8860405 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200007467.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000335.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZC3H7B

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

viral process
protein binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22006448.hg.1 ENST00000351589.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22006448.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22006448.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22006448.hg.1 TC22000335.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22006447.hg.1 ENST00000351589.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22006447.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22006447.hg.1 NM_017590.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22006447.hg.1 TC22000335.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 ENST00000486331.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay TC2200007467.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay PSR2200152793.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200152793.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay PSR2200152793.hg.1 ENST00000352645.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200152793.hg.1 NM_017590.5 Refseq
Affymetrix Human Clariom D Assay PSR2200152793.hg.1 TC2200007467.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0