This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01595310_g1
See other DUOXA2 GE Assays ›
Gene Symbol:
DUOXA2
Gene Name:
dual oxidase maturation factor 2
Gene Aliases:
SIMNIPHOM, TDH5
Chromosome Location:
Chr.15: 45114321 - 45118106 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01595310_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUOXA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207581.3 NP_997464.2 3 - 4 621 160
GenBank mRNA AK303955.1 - 3 - 4 620 160
BC137465.1 - 3 - 4 542 9021465 160
BX537581.1 - 3 - 4 905 160
DQ489734.1 - 3 - 4 472 160

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500007109.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15000351.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DUOXA2

Panther Classification:

Molecular Function -

oxidase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s)

protein transport
hydrogen peroxide metabolic process
regulation of inflammatory response
regulation of thyroid hormone generation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0