This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01595308_m1
See other DUOXA2 GE Assays ›
Gene Symbol:
DUOXA2
Gene Name:
dual oxidase maturation factor 2
Gene Aliases:
SIMNIPHOM, TDH5
Chromosome Location:
Chr.15: 45114321 - 45118106 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01595308_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUOXA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BX537581.1 - 1 - 2 283 144

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000323030.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000350243.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 ENST00000491993.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007109.hg.1 NM_207581.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0