This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01594826_m1
See other LOC400794 GE Assays ›
Gene Symbol:
LOC400794
Gene Name:
uncharacterized LOC400794
Gene Aliases:
-
Chromosome Location:
Chr.1: 165476842 - 165582155 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01594826_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC400794
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC030596.2 - 6 - 7 1341 178

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream retinoid X receptor gamma RXRG NR2B3,
RXRC
-53.7kb Sense Chr. 1 - 165400922 - 165445355 on NCBI Build GRCh38
Downstream leucine rich repeat containing 52 LRRC52 42.6kb Anti-sense Chr. 1 - 165544241 - 165563948 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0