This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01588587_g1
See other KRT14 GE Assays ›
Gene Symbol:
KRT14
Gene Name:
keratin 14
Gene Aliases:
CK14, EBS3, EBS4, K14, NFJ
Chromosome Location:
Chr.17: 41582279 - 41586895 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01588587_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KRT14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000526.4 NP_000517.2 7 - 8 1382 88
GenBank mRNA BC002690.2 - 7 - 8 1367 3610489 88
BC019097.2 - 7 - 8 1373 5013623 88
BC042437.1 - 7 - 8 1337 4776763 88
BC094830.1 - 7 - 8 1373 5443153 88
BT007186.1 - 7 - 8 1321 88
M28646.1 - 3 - 4 346 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KRT14

Gene Ontology Categories:

Function(s) Process(es)

aging
epidermis development
response to zinc ion
response to ionizing radiation
epithelial cell differentiation
hemidesmosome assembly
hair cycle
intermediate filament bundle assembly
structural constituent of cytoskeleton
protein binding
keratin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000417510.5 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000425692.6 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000583748.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009909.hg.1 ENST00000584481.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000167586.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000441550.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 ENST00000476662.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq
Affymetrix Human Clariom D Assay TC1700010680.hg.1 NM_000526.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0