This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01587093_s1
See other LOC107986876 GE Assays ›
Gene Symbol:
LOC107986876
Gene Name:
proline-rich proteoglycan 2-like
Gene Aliases:
-
Chromosome Location:
Chr.8: 22602708 - 22605328 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01587093_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC107986876
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017014122.1 XP_016869611.1 1 1067 153
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG773387.1 - 1 269 Chr. 8 - 22603774 - 22604526 [-] on Build GRCh38 153
GenBankmRNA BC031867.1 - 1 716 Chr. 8 - 22603133 - 22604966 [-] on Build GRCh38 153

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chromosome 8 open reading frame 58 C8orf58 Anti-sense Chr. 8 - 22599601 - 22604150 on NCBI Build GRCh38
Downstream bridging integrator 3 BIN3 15.5kb Sense Chr. 8 - 22620418 - 22669148 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0