Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005705.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005705.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005705.hg.1 |
TC14000456.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005707.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005707.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005707.hg.1 |
TC14000456.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005706.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005706.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005706.hg.1 |
TC14000456.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400154138.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154138.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154138.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154138.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154138.hg.1 |
TC1400007671.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154139.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154139.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154139.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154139.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154139.hg.1 |
TC1400007671.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154140.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154140.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154140.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154140.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154140.hg.1 |
TC1400007671.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007671.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154141.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154141.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154141.hg.1 |
ENST00000261980.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154141.hg.1 |
NM_182894.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154141.hg.1 |
TC1400007671.hg.1 |
Clariom D TC |