This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01573794_g1
See other SLC5A1 GE Assays ›
Gene Symbol:
SLC5A1
Gene Name:
solute carrier family 5 member 1
Gene Aliases:
D22S675, NAGT, SGLT1
Chromosome Location:
Chr.22: 32043032 - 32113029 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01573794_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC5A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000343.3 NP_000334.1 4 - 5 619 62
NM_001256314.1 NP_001243243.1 3 - 4 337 62
XM_011530331.1 XP_011528633.1 4 - 5 601 62
GenBank mRNA AK297665.1 - 3 - 4 311 62
AK312948.1 - 4 - 5 523 62
AK316184.1 - 3 - 4 316 62
BC139839.1 - 4 - 5 388 62
CR456579.1 - 4 - 5 419 62
M24847.1 - 4 - 5 379 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200007120.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000237.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK297665.1,AK316184.1,NM_001256314.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC5A1

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

intestinal D-glucose absorption
sodium ion transport
hexose transport
glucose transport
glucose transmembrane transport
glucose:sodium symporter activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22004564.hg.1 ENST00000266088.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22004564.hg.1 ENST00000543737.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22004564.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22004564.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22004564.hg.1 TC22000237.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay PSR2200150171.hg.1 TC2200007120.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0