This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01573098_g1
See other ARFGEF2 GE Assays ›
Gene Symbol:
ARFGEF2
Gene Name:
ADP ribosylation factor guanine nucleotide exchange factor 2
Gene Aliases:
BIG2, PVNH2, dJ1164I10.1
Chromosome Location:
Chr.20: 48921721 - 49036693 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01573098_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARFGEF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006420.2 NP_006411.2 15 - 16 2219 83
XM_005260252.3 XP_005260309.1 15 - 16 2233 83
GenBank mRNA AF084521.1 - 15 - 16 2067 83
BC050449.1 - 15 - 16 2219 83
BC144625.1 - 15 - 16 2130 83
BC144626.1 - 15 - 16 2130 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007620.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000382.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARFGEF2

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

receptor recycling
exocytosis
Golgi to plasma membrane transport
endosome organization
endomembrane system organization
protein transport
regulation of ARF protein signal transduction
positive regulation of tumor necrosis factor production
intracellular signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
ARF guanyl-nucleotide exchange factor activity
protein binding
myosin binding
protein kinase A regulatory subunit binding
GABA receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20005500.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20005500.hg.1 ENST00000538445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20005500.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005500.hg.1 TC20000382.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR20005502.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20005502.hg.1 ENST00000538445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20005502.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005502.hg.1 TC20000382.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000493140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000493140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000493140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000493140.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000493140.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000493140.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay PSR2000153112.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000153112.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay PSR2000153112.hg.1 ENST00000371917.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000153112.hg.1 NM_006420.2 Refseq
Affymetrix Human Clariom D Assay PSR2000153112.hg.1 TC2000007620.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0