Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020875.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020875.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020875.hg.1 |
TC08001595.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020872.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020872.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020872.hg.1 |
TC08001595.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020874.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020874.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020874.hg.1 |
TC08001595.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020873.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020873.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020873.hg.1 |
TC08001595.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011697.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179427.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179427.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179427.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179427.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179427.hg.1 |
TC0800011697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800179428.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179428.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179428.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179428.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179428.hg.1 |
TC0800011697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800179426.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179426.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179426.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179426.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179426.hg.1 |
TC0800011697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800179429.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179429.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179429.hg.1 |
ENST00000518567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179429.hg.1 |
NR_040044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179429.hg.1 |
TC0800011697.hg.1 |
Clariom D TC |