This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01568160_m1
See other ATG5 GE Assays ›
Gene Symbol:
ATG5
Gene Name:
autophagy related 5
Gene Aliases:
APG5, APG5-LIKE, APG5L, ASP, hAPG5
Chromosome Location:
Chr.6: 106184476 - 106325820 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01568160_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATG5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286106.1 NP_001273035.1 2 - 3 332 154
NM_001286108.1 NP_001273037.1 2 - 3 467 154
NM_004849.3 NP_004840.1 2 - 3 467 154
NR_104402.1 1 - 2 172 154
GenBank mRNA AK001899.1 - 2 - 3 434 154
AK315800.1 - 2 - 3 305 154
BC002699.2 - 2 - 3 313 3609927 154
BC093011.1 - 2 - 3 394 30345897 154
EU283338.1 - 1 - 2 113 154
EU283339.1 - 1 - 2 114 154
HY168998.1 - 2 - 3 335 154
JQ918353.1 - 1 - 2 114 154
JQ924061.1 - 1 - 2 114 154
Y11588.1 - 2 - 3 440 154

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600012731.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001982.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATG5

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

autophagosome assembly
mitophagy
blood vessel remodeling
regulation of cytokine secretion involved in immune response
C-terminal protein lipidation
autophagy
apoptotic process
cellular response to nitrogen starvation
response to fungus
macroautophagy
cellular homeostasis
antigen processing and presentation of endogenous antigen
negative regulation of protein ubiquitination
aggrephagy
negative stranded viral RNA replication
vasodilation
response to drug
negative regulation of apoptotic process
post-translational protein modification
nucleophagy
negative thymic T cell selection
innate immune response
otolith development
regulation of release of sequestered calcium ion into cytosol
ventricular cardiac muscle cell development
heart contraction
negative regulation of cell death
protein lipidation involved in autophagosome assembly
positive regulation of mucus secretion
regulation of cilium assembly
positive regulation of mitophagy
negative regulation of reactive oxygen species metabolic process
negative regulation of histone H4-K16 acetylation
protein binding
Atg8 ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0