This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01567791_g1
See other ANAPC4 GE Assays ›
Gene Symbol:
ANAPC4
Gene Name:
anaphase promoting complex subunit 4
Gene Aliases:
APC4
Chromosome Location:
Chr.4: 25377226 - 25418498 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01567791_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ANAPC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286756.1 NP_001273685.1 19 - 20 1511 136
XM_005248159.2 XP_005248216.1 6 - 7 405 136
XM_011513838.1 XP_011512140.1 19 - 20 1388 136
XM_011513839.1 XP_011512141.1 19 - 20 1432 136
GenBank mRNA AK023597.1 - 16 - 17 1122 136

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitotic nuclear division
regulation of mitotic metaphase/anaphase transition
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
protein K11-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
protein phosphatase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0