This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01567015_m1
See other MBNL1 GE Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01567015_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MBNL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001314057.1 NP_001300986.1 8 - 9 1648 65
NM_021038.4 NP_066368.2 6 - 7 2379 65
NM_207295.1 NP_997178.1 4 - 5 1550 65
NM_207297.1 NP_997180.1 5 - 6 1754 65
XM_005247457.4 XP_005247514.1 8 - 9 2001 65
XM_005247458.4 XP_005247515.1 8 - 9 3255 65
XM_005247460.4 XP_005247517.1 8 - 9 1964 65
XM_005247462.4 XP_005247519.1 7 - 8 1947 65
XM_005247464.4 XP_005247521.1 7 - 8 2059 65
XM_005247465.4 XP_005247522.1 8 - 9 2001 65
XM_005247471.4 XP_005247528.1 6 - 7 1977 65
XM_005247474.3 XP_005247531.1 6 - 7 919 65
XM_005247475.3 XP_005247532.1 7 - 8 1242 65
XM_005247476.4 XP_005247533.1 5 - 6 955 65
XM_005247477.4 XP_005247534.1 6 - 7 1008 65
XM_006713639.1 XP_006713702.1 8 - 9 1983 65
XM_011512843.2 XP_011511145.1 7 - 8 1797 65
XM_011512845.2 XP_011511147.1 6 - 7 1743 65
XM_011512847.2 XP_011511149.1 5 - 6 1706 65
XM_011512848.2 XP_011511150.1 8 - 9 1605 65
XM_017006423.1 XP_016861912.1 7 - 8 2252 65
XM_017006424.1 XP_016861913.1 7 - 8 2079 65
XM_017006427.1 XP_016861916.1 6 - 7 2172 65
XM_017006428.1 XP_016861917.1 6 - 7 1836 65
XM_017006429.1 XP_016861918.1 6 - 7 1919 65
XM_017006432.1 XP_016861921.1 7 - 8 2036 65
XM_017006435.1 XP_016861924.1 7 - 8 1049 65
XM_017006436.1 XP_016861925.1 6 - 7 3108 65
XM_017006439.1 XP_016861928.1 6 - 7 1188 65
XM_017006440.1 XP_016861929.1 5 - 6 922 65
GenBank mRNA AF395876.1 - 4 - 5 761 65
AF401998.1 - 5 - 6 965 65
AF497719.1 - 5 - 6 965 65
AJ308400.1 - 5 - 6 994 65
AK304089.1 - 5 - 6 728 65
AK307420.1 - 4 - 5 569 65
BC043493.1 - 6 - 7 1884 6044671 65
BC050535.1 - 8 - 9 1169 6042311 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MBNL1

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0