This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01562425_m1
See other PMS2 GE Assays ›
Gene Symbol:
PMS2
Gene Name:
PMS1 homolog 2, mismatch repair system component
Gene Aliases:
HNPCC4, MLH4, PMS2CL, PMSL2
Chromosome Location:
Chr.7: 5970925 - 6009106 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01562425_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PMS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000535.6 NP_000526.2 5 - 6 625 92
NM_001322003.1 NP_001308932.1 5 - 6 620 92
NM_001322004.1 NP_001308933.1 4 - 5 485 92
NM_001322005.1 NP_001308934.1 5 - 6 737 92
NM_001322006.1 NP_001308935.1 5 - 6 625 92
NM_001322007.1 NP_001308936.1 4 - 5 522 92
NM_001322008.1 NP_001308937.1 3 - 4 382 92
NM_001322009.1 NP_001308938.1 5 - 6 732 92
NM_001322010.1 NP_001308939.1 4 - 5 485 92
NM_001322014.1 NP_001308943.1 5 - 6 625 92
NM_001322015.1 NP_001308944.1 5 - 6 795 92
NR_136154.1 5 - 6 625 92
GenBank mRNA AB103082.1 - 5 - 6 538 92
AB103083.1 - 5 - 6 538 92
AB103086.1 - 2 - 3 220 92
AK294661.1 - 4 - 5 475 92
AK312390.1 - 5 - 6 574 92
BC008400.1 - 5 - 6 583 92
BC031832.1 - 5 - 6 589 92
BC093921.1 - 5 - 6 560 7939766 92
BP251389.1 - 1 - 2 95 92
CR981250.1 - 5 - 6 526 92
U14658.1 - 5 - 6 540 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700010182.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001124.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PMS2

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mismatch repair
somatic hypermutation of immunoglobulin genes
response to drug
nucleic acid phosphodiester bond hydrolysis
DNA binding
single-stranded DNA binding
endonuclease activity
protein binding
ATP binding
ATPase activity
single base insertion or deletion binding
MutSalpha complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000469652.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000469652.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0