This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01561010_m1
See other FKBP5 GE Assays ›
Gene Symbol:
FKBP5
Gene Name:
FK506 binding protein 5
Gene Aliases:
AIG6, FKBP51, FKBP54, P54, PPIase, Ptg-10
Chromosome Location:
Chr.6: 35573585 - 35728583 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01561010_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FKBP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145775.2 NP_001139247.1 10 - 11 1336 89
NM_001145776.1 NP_001139248.1 9 - 10 1223 89
NM_004117.3 NP_004108.1 9 - 10 1178 89
GenBank mRNA AF194172.1 - 9 - 10 1173 89
AK222807.1 - 9 - 10 1151 89
AK302704.1 - 5 - 6 624 89
AK303356.1 - 9 - 10 1040 89
AK312422.1 - 9 - 10 1223 89
BC042605.1 - 10 - 11 1336 4539929 89
BC111050.1 - 9 - 10 1139 5723416 89
BX647390.1 - 9 - 10 1151 89
U42031.1 - 8 - 9 997 89
U71321.1 - 9 - 10 1173 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011635.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06004150.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FKBP5

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
chaperone-mediated protein folding
peptidyl-prolyl cis-trans isomerase activity
protein binding
FK506 binding
heat shock protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000357266.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000452048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000452048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000452048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000452048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000536438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000539068.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 ENST00000542713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145775.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145776.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_001145777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NM_004117.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011635.hg.1 NR_027117.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0