Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048933.hg.1 |
ENST00000331128.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048933.hg.1 |
NM_175065.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048933.hg.1 |
TC01003194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015711.hg.1 |
ENST00000331128.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015711.hg.1 |
NM_175065.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015711.hg.1 |
NM_175065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015711.hg.1 |
ENST00000331128.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015711.hg.1 |
NM_175065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015711.hg.1 |
NM_175065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217614.hg.1 |
ENST00000331128.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217614.hg.1 |
NM_175065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217614.hg.1 |
ENST00000331128.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217614.hg.1 |
NM_175065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217614.hg.1 |
TC0100015711.hg.1 |
Clariom D TC |