This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01560181_g1
See other EMC3 GE Assays ›
Gene Symbol:
EMC3
Gene Name:
ER membrane protein complex subunit 3
Gene Aliases:
POB, TMEM111
Chromosome Location:
Chr.3: 9962680 - 10011248 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01560181_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EMC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018447.3 NP_060917.1 2 - 3 615 65
XM_005265321.3 XP_005265378.1 2 - 3 483 65
GenBank mRNA AF157321.1 - 1 - 2 327 65
AK172843.1 - 1 - 2 323 65
AK312008.1 - 1 - 2 322 65
BC022807.1 - 1 - 2 310 4897577 65
DA359966.1 - 2 - 3 483 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013934.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03003327.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EMC3

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein folding in endoplasmic reticulum
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03019954.hg.1 ENST00000245046.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019954.hg.1 ENST00000429759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019954.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019954.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03019954.hg.1 TC03003327.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03019955.hg.1 ENST00000245046.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019955.hg.1 ENST00000429759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019955.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019955.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03019955.hg.1 TC03003327.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03019959.hg.1 ENST00000245046.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019959.hg.1 ENST00000429759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019959.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03019959.hg.1 TC03003327.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03019960.hg.1 ENST00000245046.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019960.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03019960.hg.1 TC03003327.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03019953.hg.1 ENST00000245046.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019953.hg.1 ENST00000429759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019953.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019953.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03019953.hg.1 TC03003327.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03019956.hg.1 ENST00000245046.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019956.hg.1 ENST00000429759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019956.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03019956.hg.1 NM_018447.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03019956.hg.1 TC03003327.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay TC0300013934.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206551.hg.1 TC0300013934.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300206557.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206557.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206557.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206557.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206557.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206557.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206557.hg.1 TC0300013934.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206549.hg.1 TC0300013934.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206550.hg.1 TC0300013934.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 ENST00000497557.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206552.hg.1 TC0300013934.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 ENST00000245046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 ENST00000429759.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 ENST00000487465.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 NM_018447.3 Refseq
Affymetrix Human Clariom D Assay PSR0300206555.hg.1 TC0300013934.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 193395 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0