This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01559366_m1
See other UNC5D GE Assays ›
Gene Symbol:
UNC5D
Gene Name:
unc-5 netrin receptor D
Gene Aliases:
PRO34692, Unc5h4
Chromosome Location:
Chr.8: 35235457 - 35796550 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01559366_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC5D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322818.1 NP_001309747.1 11 - 12 1814 89
NM_080872.3 NP_543148.2 11 - 12 2101 89
XM_011544400.2 XP_011542702.1 10 - 11 2042 89
XM_011544401.2 XP_011542703.1 11 - 12 2036 89
XM_011544402.2 XP_011542704.1 10 - 11 1907 89
XM_011544403.2 XP_011542705.1 9 - 10 1874 89
XM_011544404.2 XP_011542706.1 10 - 11 1868 89
XM_017013044.1 XP_016868533.1 12 - 13 2148 89
XM_017013045.1 XP_016868534.1 12 - 13 2118 89
XM_017013046.1 XP_016868535.1 11 - 12 2115 89
XM_017013047.1 XP_016868536.1 11 - 12 1979 89
XM_017013048.1 XP_016868537.1 11 - 12 3261 89
XM_017013049.1 XP_016868538.1 11 - 12 3212 89
XM_017013050.1 XP_016868539.1 12 - 13 2668 89
GenBank mRNA AB055056.1 - 11 - 12 1813 89
AK122610.1 - 3 - 4 612 89
AY358147.1 - 11 - 12 2101 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800007269.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000266.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UNC5D

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

apoptotic process
signal transduction
axon guidance
pyramidal neuron differentiation
cell-cell adhesion
regulation of neuron migration

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000287272.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000404895.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000416672.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000420357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000449677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000453357.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000474634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000474634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000474634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000474634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000474634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000474634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 ENST00000474634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007269.hg.1 NM_080872.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0