This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01554355_m1
See other GZMB GE Assays ›
Gene Symbol:
GZMB
Gene Name:
granzyme B
Gene Aliases:
CCPI, CGL-1, CGL1, CSP-B, CSPB, CTLA1, CTSGL1, HLP, SECT
Chromosome Location:
Chr.14: 24630954 - 24634226 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01554355_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GZMB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004131.4 NP_004122.2 1 - 2 123 134
GenBank mRNA AY232655.1 - 1 - 2 60 134
AY372494.1 - 1 - 2 164 134
BC030195.1 - 1 - 2 79 5223876 134
BX283601.1 - 1 - 2 125 134
J03189.1 - 1 - 2 96 134
J04071.1 - 1 - 2 73 134
M17016.1 - 1 - 2 64 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400008764.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000982.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protease Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GZMB

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
protein processing
cytolysis
natural killer cell mediated cytotoxicity
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
serine-type endopeptidase activity
protein binding
serine-type peptidase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14011272.hg.1 ENST00000216341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011272.hg.1 ENST00000382540.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011272.hg.1 ENST00000382542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011272.hg.1 ENST00000415355.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011272.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011272.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14011272.hg.1 TC14000982.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14011275.hg.1 ENST00000216341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011275.hg.1 ENST00000382539.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011275.hg.1 ENST00000382540.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011275.hg.1 ENST00000382542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011275.hg.1 ENST00000415355.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011275.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14011275.hg.1 NM_004131.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14011275.hg.1 TC14000982.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay PSR1400164593.hg.1 TC1400008764.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000532263.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 ENST00000616551.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay TC1400008764.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000216341.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000382540.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000382542.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000415355.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000526004.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000530830.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 ENST00000554242.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 NM_004131.5 Refseq
Affymetrix Human Clariom D Assay PSR1400164589.hg.1 TC1400008764.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 197238 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0