This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01553226_m1
See other PPP6R1 GE Assays ›
Gene Symbol:
PPP6R1
Gene Name:
protein phosphatase 6 regulatory subunit 1
Gene Aliases:
KIAA1115, PP6R1, SAP190, SAPS1
Chromosome Location:
Chr.19: 55229779 - 55258670 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01553226_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP6R1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK297152.1 - 1 - 2 94 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK297152.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000586690.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000586690.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000586690.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000586690.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000589343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000589343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000589343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000591602.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000592242.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000592242.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000592242.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 ENST00000592242.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000412770.6 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000586690.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000586690.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000586690.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000586690.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587283.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000587457.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000589343.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000589343.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000589343.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591323.5 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591602.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591602.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591602.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000591602.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000592242.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000592242.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000592242.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 ENST00000592242.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq
Affymetrix Human Clariom D Assay TC1900012050.hg.1 NM_014931.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0