This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01552083_m1
See other ALDH1A3 GE Assays ›
Gene Symbol:
ALDH1A3
Gene Name:
aldehyde dehydrogenase 1 family member A3
Gene Aliases:
ALDH1A6, ALDH6, MCOP8, RALDH3
Chromosome Location:
Chr.15: 100879692 - 100916626 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01552083_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH1A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000693.3 NP_000684.2 12 - 13 1683 79
NM_001293815.1 NP_001280744.1 9 - 10 1362 79
GenBank mRNA AK302607.1 - 11 - 12 1304 79
AK312303.1 - 12 - 13 1544 79
BC069274.1 - 12 - 13 1523 6208628 79
BX538027.1 - 9 - 10 1180 79
U07919.1 - 12 - 13 1519 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500010800.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ALDH1A3

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

optic cup morphogenesis involved in camera-type eye development
retinoic acid biosynthetic process
locomotory behavior
nucleus accumbens development
inner ear morphogenesis
retinol metabolic process
retinoic acid metabolic process
retinal metabolic process
positive regulation of apoptotic process
embryonic eye morphogenesis
neuromuscular process controlling balance
oxidation-reduction process
righting reflex
olfactory pit development
face development
retinal dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
aldehyde dehydrogenase [NAD(P)+] activity
protein homodimerization activity
thyroid hormone binding
NAD+ binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000557963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000557963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000557963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000558869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay PSR1500183824.hg.1 TC1500010800.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay PSR1500183825.hg.1 TC1500010800.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000329841.9 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000346623.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000557963.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000557963.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000557963.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558033.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558869.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558869.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558869.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000558869.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000560555.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 ENST00000561338.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_000693.3 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq
Affymetrix Human Clariom D Assay TC1500010800.hg.1 NM_001293815.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header