Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011767.hg.1 |
ENST00000290650.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011767.hg.1 |
ENST00000382177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011767.hg.1 |
NM_174916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011767.hg.1 |
TC15001271.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500166990.hg.1 |
ENST00000290650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500166990.hg.1 |
NM_174916.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500166990.hg.1 |
ENST00000290650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500166990.hg.1 |
NM_174916.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500166990.hg.1 |
TC1500009204.hg.1 |
Clariom D TC |