This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01550696_m1
See other SCCPDH GE Assays ›
Gene Symbol:
SCCPDH
Gene Name:
saccharopine dehydrogenase (putative)
Gene Aliases:
CGI-49, NET11
Chromosome Location:
Chr.1: 246724076 - 246768137 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01550696_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCCPDH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016002.2 NP_057086.2 10 - 11 1449 104
GenBank mRNA AF151807.1 - 10 - 11 1099 104
AK075178.1 - 10 - 11 1205 104
BC026185.2 - 10 - 11 1110 4798194 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100012278.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002003.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SCCPDH

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01030976.hg.1 ENST00000366509.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01030976.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01030976.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01030976.hg.1 TC01002003.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay TC0100012278.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay PSR0100190312.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100190312.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay PSR0100190312.hg.1 ENST00000366510.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100190312.hg.1 NM_016002.2 Refseq
Affymetrix Human Clariom D Assay PSR0100190312.hg.1 TC0100012278.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0