Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
ENST00000356219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
ENST00000397355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
ENST00000404121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
ENST00000404621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
ENST00000405201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
ENST00000429285.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
ENST00000447011.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
NM_001077261.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
NM_001206654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
NM_006312.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027724.hg.1 |
TC12002096.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000356219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000404121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000404621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000405201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000429285.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000448614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000458234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
NM_001077261.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
NM_001206654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
NM_006312.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000356219.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000404121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000404621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000405201.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000429285.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000448614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
ENST00000458234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
NM_001077261.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
NM_001206654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
NM_006312.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196458.hg.1 |
TC1200012308.hg.1 |
Clariom D TC |