This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01547992_m1
See other ANK2 GE Assays ›
Gene Symbol:
ANK2
Gene Name:
ankyrin 2, neuronal
Gene Aliases:
ANK-2, LQT4, brank-2
Chromosome Location:
Chr.4: 112706083 - 113383740 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01547992_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ANK2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127493.1 NP_001120965.1 39 - 40 4621 76
NM_020977.3 NP_066187.2 37 - 38 4527 76
XM_017008078.1 XP_016863567.1 37 - 38 4625 76
XM_017008079.1 XP_016863568.1 35 - 36 4355 76
XM_017008080.1 XP_016863569.1 37 - 38 4476 76
XM_017008081.1 XP_016863570.1 36 - 37 4526 76
XM_017008082.1 XP_016863571.1 38 - 39 4703 76
XM_017008083.1 XP_016863572.1 38 - 39 4766 76
XM_017008084.1 XP_016863573.1 38 - 39 4661 76
XM_017008085.1 XP_016863574.1 38 - 39 4622 76
XM_017008086.1 XP_016863575.1 37 - 38 4523 76
XM_017008087.1 XP_016863576.1 39 - 40 4802 76
XM_017008088.1 XP_016863577.1 40 - 41 4596 76
XM_017008089.1 XP_016863578.1 37 - 38 4769 76
XM_017008090.1 XP_016863579.1 38 - 39 4766 76
XM_017008091.1 XP_016863580.1 36 - 37 4463 76
XM_017008092.1 XP_016863581.1 38 - 39 4563 76
XM_017008093.1 XP_016863582.1 39 - 40 4560 76
XM_017008094.1 XP_016863583.1 38 - 39 4622 76
XM_017008095.1 XP_016863584.1 36 - 37 4440 76
XM_017008096.1 XP_016863585.1 40 - 41 4596 76
XM_017008097.1 XP_016863586.1 37 - 38 4401 76
XM_017008098.1 XP_016863587.1 38 - 39 4563 76
XM_017008099.1 XP_016863588.1 39 - 40 4560 76
XM_017008100.1 XP_016863589.1 36 - 37 4365 76
XM_017008101.1 XP_016863590.1 36 - 37 4526 76
XM_017008102.1 XP_016863591.1 35 - 36 4341 76
XM_017008103.1 XP_016863592.1 38 - 39 4622 76
XM_017008104.1 XP_016863593.1 37 - 38 4464 76
XM_017008105.1 XP_016863594.1 36 - 37 4440 76
XM_017008106.1 XP_016863595.1 35 - 36 4427 76
XM_017008107.1 XP_016863596.1 37 - 38 4401 76
XM_017008108.1 XP_016863597.1 35 - 36 4325 76
XM_017008110.1 XP_016863599.1 37 - 38 4464 76
XM_017008111.1 XP_016863600.1 36 - 37 4440 76
XM_017008112.1 XP_016863601.1 34 - 35 4242 76
GenBank mRNA AK294720.1 - 16 - 17 2272 76
AK299893.1 - 10 - 11 1369 76
BC125235.1 - 5 - 6 412 76
BC125236.1 - 5 - 6 412 76
BX537758.1 - 39 - 40 4595 76
X56958.2 - 37 - 38 4442 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400008450.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000584.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ANK2

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of heart rate
atrial septum development
cellular calcium ion homeostasis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
positive regulation of gene expression
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of cardiac muscle contraction by calcium ion signaling
paranodal junction assembly
regulation of protein stability
T-tubule organization
protein localization to organelle
protein localization to cell surface
cellular protein localization
protein localization to M-band
protein localization to T-tubule
positive regulation of potassium ion transport
protein stabilization
regulation of release of sequestered calcium ion into cytosol
response to methylmercury
regulation of calcium ion transport
positive regulation of calcium ion transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
sarcoplasmic reticulum calcium ion transport
protein localization to endoplasmic reticulum
protein localization to plasma membrane
protein targeting to plasma membrane
regulation of cardiac muscle cell contraction
ventricular cardiac muscle cell action potential
atrial cardiac muscle cell action potential
SA node cell action potential
membrane depolarization during SA node cell action potential
atrial cardiac muscle cell to AV node cell communication
SA node cell to atrial cardiac muscle cell communication
regulation of heart rate by cardiac conduction
regulation of SA node cell action potential
regulation of atrial cardiac muscle cell action potential
positive regulation of potassium ion transmembrane transporter activity
regulation of calcium ion transmembrane transporter activity
positive regulation of calcium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
protein kinase binding
spectrin binding
protein binding, bridging
ion channel binding
ATPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0