This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01547829_m1
See other CA6 GE Assays ›
Gene Symbol:
CA6
Gene Name:
carbonic anhydrase 6
Gene Aliases:
CA-VI, GUSTIN
Chromosome Location:
Chr.1: 8945786 - 8975092 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01547829_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CA6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001215.3 NP_001206.2 3 - 4 455 90
NM_001270500.1 NP_001257429.1 3 - 4 455 90
NM_001270501.1 NP_001257430.1 2 - 3 275 90
XM_011542083.2 XP_011540385.1 3 - 4 450 90
XM_011542084.2 XP_011540386.1 3 - 4 503 90
GenBank mRNA AB102863.1 - 3 - 4 402 90
AB103091.1 - 2 - 3 222 90
AK300652.1 - 3 - 4 419 90
BP333541.1 - 3 - 4 454 90
BP334640.1 - 3 - 4 425 90
M57892.1 - 3 - 4 408 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100006761.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000110.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CA6

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
one-carbon metabolic process
bicarbonate transport
carbonate dehydratase activity
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01001719.hg.1 ENST00000377436.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001719.hg.1 ENST00000377442.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001719.hg.1 ENST00000377443.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001719.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001719.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001719.hg.1 TC01000110.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01001718.hg.1 ENST00000377436.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001718.hg.1 ENST00000377442.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001718.hg.1 ENST00000377443.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01001718.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01001718.hg.1 TC01000110.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay PSR0100146768.hg.1 TC0100006761.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377436.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377442.3 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000377443.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000476083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000480186.7 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 ENST00000549778.5 Ensembl
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001215.3 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270500.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270501.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq
Affymetrix Human Clariom D Assay TC0100006761.hg.1 NM_001270502.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header