This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01547093_m1
See other SYT12 GE Assays ›
Gene Symbol:
SYT12
Gene Name:
synaptotagmin 12
Gene Aliases:
SYT11, sytXII
Chromosome Location:
Chr.11: 67006778 - 67050863 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01547093_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SYT12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177880.1 NP_001171351.1 4 - 5 850 101
NM_001318773.1 NP_001305702.1 5 - 6 975 101
NM_001318775.1 NP_001305704.1 3 - 4 565 101
NM_177963.3 NP_808878.1 4 - 5 759 101
XM_006718737.3 XP_006718800.1 6 - 7 1610 101
XM_011545346.2 XP_011543648.1 7 - 8 1782 101
XM_011545347.1 XP_011543649.1 4 - 5 700 101
XM_017018547.1 XP_016874036.1 5 - 6 863 101
XM_017018548.1 XP_016874037.1 5 - 6 791 101
GenBank mRNA AK024280.1 - 7 - 8 1782 101
AK289672.1 - 4 - 5 712 101
AK289727.1 - 4 - 5 712 101
AK315810.1 - 4 - 5 851 101
AL833834.1 - 2 - 3 441 101
BC037406.1 - 4 - 5 684 5770140 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100008111.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000678.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SYT12

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

vesicle fusion
regulation of calcium ion-dependent exocytosis
regulation of neurotransmitter secretion
synaptic vesicle endocytosis
calcium ion-regulated exocytosis of neurotransmitter
spontaneous exocytosis of neurotransmitter
long-term synaptic potentiation
calcium ion binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000393946.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000525457.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000526281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000526281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000526281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000526281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000526281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000526281.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000527043.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000531392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000531392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000531392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000531392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000531392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000531392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000533427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000533427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000533427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 ENST00000533427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_001177880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008111.hg.1 NM_177963.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0