This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01385224_m1
See other MYH16 GE Assays ›
Gene Symbol:
MYH16
Gene Name:
myosin heavy chain 16 pseudogene
Gene Aliases:
MHC20, MYH16P, MYH5
Chromosome Location:
Chr.7: 99273301 - 99297972 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01385224_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYH16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002147.2 13 - 14 1836 78
GenBank mRNA AK025690.1 - 13 - 14 1836 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream karyopherin subunit alpha 7 KPNA7 IPOA8 -74.7kb Anti-sense Chr. 7 - 99145467 - 99219417 on NCBI Build GRCh38
Downstream actin related protein 2/3 complex subunit 1A ARPC1A Arc40,
HEL-68,
HEL-S-307,
SOP2Hs,
SOP2L
29.0kb Sense Chr. 7 - 99325873 - 99366262 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700013417.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000591.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Process(es)

motor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07008661.hg.1 NR_002147.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07008661.hg.1 TC07000591.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0700197938.hg.1 NR_002147.2 Refseq
Affymetrix Human Clariom D Assay PSR0700197938.hg.1 NR_002147.2 Refseq
Affymetrix Human Clariom D Assay PSR0700197938.hg.1 TC0700013417.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0