This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01376457_m1
See other CEBPZOS GE Assays ›
Gene Symbol:
CEBPZOS
Gene Name:
CEBPZ opposite strand
Gene Aliases:
CEBPZ-AS1
Chromosome Location:
Chr.2: 37196451 - 37216193 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01376457_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEBPZOS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322373.1 NP_001309302.1 2 - 3 242 140
XM_017003106.1 XP_016858595.1 2 - 3 271 140
GenBank mRNA BC017652.1 - 1 - 2 169 140

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001322373.1, XM_017003106.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000392061.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000392061.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000392061.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000392061.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000470216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000470216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000470216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016428.hg.1 ENST00000470216.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000392061.6 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000392061.6 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000392061.6 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000392061.6 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000397226.2 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000402297.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000406711.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000470216.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000470216.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000470216.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016428.hg.1 ENST00000470216.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0