This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01369441_g1
See other LOC101929268 GE Assays ›
Gene Symbol:
LOC101929268
Gene Name:
uncharacterized LOC101929268
Gene Aliases:
-
Chromosome Location:
Chr.8: 48551567 - 48698510 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01369441_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101929268
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105002.1 1 - 2 94 119
GenBank mRNA AK092055.1 - 1 - 2 94 119
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA653432.1 - 1 - 2 94 Chr. 8 - 48551567 - 48696671 [+] on Build GRCh38 119

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ubiquitin conjugating enzyme E2 V2 UBE2V2 DDVIT1,
DDVit-1,
EDAF-1,
EDPF-1,
EDPF1,
MMS2,
UEV-2,
UEV2
-486.9kb Sense Chr. 8 - 47997527 - 48063545 on NCBI Build GRCh38
Downstream uncharacterized LOC101929217 LOC101929217 68.3kb Sense Chr. 8 - 48620403 - 48623895 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800007567.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay PSR0800152281.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800152281.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay PSR0800152281.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800152281.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay PSR0800152281.hg.1 TC0800007567.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007567.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay TC0800007567.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay PSR0800152282.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800152282.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay PSR0800152282.hg.1 ENST00000522575.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800152282.hg.1 NR_105002.1 Refseq
Affymetrix Human Clariom D Assay PSR0800152282.hg.1 TC0800007567.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header