Assay Details
Gene Symbol: DIAPH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046539.2 | 2 - 3 | 230 | 76 | ||
NR_051994.2 | 2 - 3 | 158 | 76 | |||
GenBank mRNA | AW468687.1 | - | 2 - 3 | 76 | ||
BX093461.1 | - | 2 - 3 | 229 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000432995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000432995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000432995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_051993.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_051993.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_051993.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007335.hg.1 | NR_109839.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000432995.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | NR_051993.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000432995.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | NR_051993.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300149683.hg.1 | TC1300007335.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000422052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000432995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000432995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000432995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | ENST00000435636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_046539.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_051993.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_051993.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_051993.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_051994.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109838.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007335.hg.1 | NR_109839.1 | Refseq |
