This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01128446_m1
See other GRPEL2 GE Assays ›
Gene Symbol:
GRPEL2
Gene Name:
GrpE like 2, mitochondrial
Gene Aliases:
Mt-GrpE#2
Chromosome Location:
Chr.5: 149345414 - 149354583 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01128446_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRPEL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152407.3 NP_689620.2 1 - 2 204 74
GenBank mRNA AK074293.1 - 1 - 2 90 74
AK294005.1 - 1 - 2 116 74
AL704536.1 - 1 - 2 205 74
AL832325.1 - 1 - 2 99 74
BC036678.1 - 1 - 2 124 5259464 74
BC070090.1 - 1 - 2 106 30347705 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500009059.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000818.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GRPEL2

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

protein folding
regulation of catalytic activity
adenyl-nucleotide exchange factor activity
protein homodimerization activity
unfolded protein binding
chaperone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00009885.hg.1 ENST00000421856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008372.hg.1 ENST00000448626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011688.hg.1 ENST00000329271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011688.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011688.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011688.hg.1 ENST00000513661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011688.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05011688.hg.1 TC05000818.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05011689.hg.1 ENST00000329271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011689.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011689.hg.1 ENST00000513661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011689.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05011689.hg.1 TC05000818.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05011691.hg.1 ENST00000329271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011691.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011691.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011691.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05011691.hg.1 TC05000818.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05011686.hg.1 ENST00000329271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011686.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011686.hg.1 ENST00000513661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011686.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05011686.hg.1 TC05000818.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05011683.hg.1 ENST00000329271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011683.hg.1 ENST00000513661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05011683.hg.1 NM_152407.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05011683.hg.1 TC05000818.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay TC0500009059.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163165.hg.1 TC0500009059.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163160.hg.1 TC0500009059.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163162.hg.1 TC0500009059.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163163.hg.1 TC0500009059.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500163156.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163156.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163156.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163156.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163156.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163156.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163156.hg.1 TC0500009059.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 ENST00000513661.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163159.hg.1 TC0500009059.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700008372.hg.1 ENST00000448626.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009885.hg.1 ENST00000421856.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 ENST00000329271.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 ENST00000416916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 ENST00000507562.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 NM_152407.3 Refseq
Affymetrix Human Clariom D Assay PSR0500163166.hg.1 TC0500009059.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0