Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017484.hg.1 |
ENST00000296589.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017484.hg.1 |
ENST00000342059.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017484.hg.1 |
ENST00000345083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017484.hg.1 |
ENST00000382102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017484.hg.1 |
ENST00000509381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017484.hg.1 |
NM_001012509.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017484.hg.1 |
NM_016180.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017484.hg.1 |
TC05001245.hg.1 |
HTA TC |