This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01127782_m1
See other PDE6C GE Assays ›
Gene Symbol:
PDE6C
Gene Name:
phosphodiesterase 6C
Gene Aliases:
ACHM5, COD4, PDEA2
Chromosome Location:
Chr.10: 93612588 - 93665673 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01127782_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE6C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006204.3 NP_006195.3 5 - 6 1076 124
XM_017016322.1 XP_016871811.1 3 - 4 440 124
GenBank mRNA U31973.1 - 5 - 6 1076 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008488.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000667.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDE6C

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

visual perception
phototransduction, visible light
retinal cone cell development
cGMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10007510.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007510.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007510.hg.1 TC10000667.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay PSR1000157712.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000157712.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay PSR1000157712.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000157712.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay PSR1000157712.hg.1 TC1000008488.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0