This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01127710_m1
See other ROCK1 GE Assays ›
Gene Symbol:
ROCK1
Gene Name:
Rho associated coiled-coil containing protein kinase 1
Gene Aliases:
P160ROCK, ROCK-I
Chromosome Location:
Chr.18: 20949740 - 21111851 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01127710_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROCK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005406.2 NP_005397.1 2 - 3 1120 164
GenBank mRNA AK057038.1 - 2 - 3 1121 164
AK307388.1 - 2 - 3 935 164
BC113114.1 - 2 - 3 195 7961989 164
U43195.1 - 2 - 3 179 164

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800008219.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000397.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ROCK1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

apical constriction
protein phosphorylation
cellular component disassembly involved in execution phase of apoptosis
smooth muscle contraction
leukocyte cell-cell adhesion
signal transduction
I-kappaB kinase/NF-kappaB signaling
Rho protein signal transduction
negative regulation of angiogenesis
membrane to membrane docking
actin cytoskeleton organization
regulation of cell adhesion
cortical actin cytoskeleton organization
bleb assembly
negative regulation of protein binding
regulation of actin cytoskeleton organization
negative regulation of myosin-light-chain-phosphatase activity
negative regulation of neuron apoptotic process
regulation of keratinocyte differentiation
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
leukocyte migration
leukocyte tethering or rolling
myoblast migration
regulation of stress fiber assembly
regulation of focal adhesion assembly
positive regulation of focal adhesion assembly
establishment of protein localization to plasma membrane
regulation of establishment of endothelial barrier
negative regulation of bicellular tight junction assembly
regulation of establishment of cell polarity
regulation of cell motility
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
GTP-Rho binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000578051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000578051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000578051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000582445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000582445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000582445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000583556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000583556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000583556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000584687.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000584687.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000584687.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000584875.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000584875.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000584875.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 ENST00000584875.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18004093.hg.1 ENST00000399799.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18004093.hg.1 NM_005406.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18004093.hg.1 TC18000397.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000578051.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000578051.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000578051.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000582445.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000582445.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000582445.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000583556.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000583556.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000583556.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000584687.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000584687.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000584687.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000584875.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000584875.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000584875.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 ENST00000584875.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay TC1800008219.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay PSR1800156157.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay PSR1800156157.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay PSR1800156157.hg.1 ENST00000399799.2 Ensembl
Affymetrix Human Clariom D Assay PSR1800156157.hg.1 NM_005406.2 Refseq
Affymetrix Human Clariom D Assay PSR1800156157.hg.1 TC1800008219.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0