This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01127517_m1
See other SDHAF2 GE Assays ›
Gene Symbol:
SDHAF2
Gene Name:
succinate dehydrogenase complex assembly factor 2
Gene Aliases:
C11orf79, PGL2, SDH5
Chromosome Location:
Chr.11: 61430125 - 61446767 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01127517_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDHAF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017841.2 NP_060311.1 1 - 2 59 81
GenBank mRNA AK000494.1 - 1 - 2 59 81
BC002331.2 - 1 - 2 42 3535474 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013018.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003455.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SDHAF2

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, succinate to ubiquinone
protein dephosphorylation
negative regulation of epithelial to mesenchymal transition
protein-FAD linkage
negative regulation of canonical Wnt signaling pathway
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000623232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 ENST00000623232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11006443.hg.1 ENST00000301761.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11006443.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11006443.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11006443.hg.1 TC11003455.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11006442.hg.1 ENST00000301761.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11006442.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11006442.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11006442.hg.1 NM_017841.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11006442.hg.1 TC11003455.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000623232.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 ENST00000623232.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay PSR1100200794.hg.1 TC1100013018.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542074.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000623232.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 ENST00000623232.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay TC1100013018.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000301761.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000359614.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000534878.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000536250.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000537782.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000542794.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 ENST00000543265.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 NM_017841.2 Refseq
Affymetrix Human Clariom D Assay PSR1100200798.hg.1 TC1100013018.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0