Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000551673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000551673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000551673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
ENST00000551673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017906.hg.1 |
ENST00000324868.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017906.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017906.hg.1 |
TC12001376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017905.hg.1 |
ENST00000324868.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017905.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017905.hg.1 |
TC12001376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017904.hg.1 |
ENST00000324868.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017904.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017904.hg.1 |
TC12001376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017907.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017907.hg.1 |
TC12001376.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200178216.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178216.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178216.hg.1 |
TC1200010336.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200178211.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178211.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178211.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178211.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178211.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178211.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178211.hg.1 |
TC1200010336.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200178212.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178212.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178212.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178212.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178212.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178212.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178212.hg.1 |
TC1200010336.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200178215.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178215.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178215.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178215.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178215.hg.1 |
TC1200010336.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000548490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000551673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000551673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000551673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
ENST00000551673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010336.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178213.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178213.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178213.hg.1 |
ENST00000324868.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200178213.hg.1 |
NM_001040436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200178213.hg.1 |
TC1200010336.hg.1 |
Clariom D TC |