Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001716.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001716.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001716.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001716.hg.1 |
TC13000147.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300147350.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147350.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147350.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147350.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147350.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147350.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147350.hg.1 |
TC1300006979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000379359.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
ENST00000487837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006979.hg.1 |
NM_014059.2 |
Refseq |