This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01120905_m1
See other BIN1 GE Assays ›
Gene Symbol:
BIN1
Gene Name:
bridging integrator 1
Gene Aliases:
AMPH2, AMPHL, SH3P9
Chromosome Location:
Chr.2: 127048023 - 127107400 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01120905_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BIN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320641.1 NP_001307570.1 11 - 12 1419 74
NM_001320642.1 NP_001307571.1 12 - 13 2006 74
NM_139343.2 NP_647593.1 12 - 13 1512 74
XM_005263642.3 XP_005263699.1 13 - 14 1630 74
XM_005263643.3 XP_005263700.1 12 - 13 1537 74
XM_005263645.2 XP_005263702.1 12 - 13 1021 74
XM_006712425.3 XP_006712488.1 13 - 14 1630 74
XM_006712427.2 XP_006712490.1 11 - 12 976 74
XM_011510975.2 XP_011509277.1 13 - 14 2263 74
XM_017003819.1 XP_016859308.1 13 - 14 1444 74
XM_017003820.1 XP_016859309.1 11 - 12 1492 74
GenBank mRNA AF004015.1 - 12 - 13 1128 74
AF070576.1 - 8 - 9 734 74
AL713697.1 - 12 - 13 1473 74
BE047931.1 - 3 - 4 317 74
U87558.1 - 12 - 13 1149 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200014160.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002276.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BIN1

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
nucleus organization
cell proliferation
viral process
regulation of endocytosis
muscle cell differentiation
positive regulation of apoptotic process
regulation of neuron differentiation
positive regulation of astrocyte differentiation
lipid tube assembly
regulation of cell cycle arrest
protein binding
identical protein binding
tau protein binding
actin filament binding
RNA polymerase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02036550.hg.1 ENST00000316724.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036550.hg.1 ENST00000357970.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036550.hg.1 NM_139343.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036550.hg.1 NM_139344.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036550.hg.1 TC02002276.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 ENST00000316724.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 ENST00000357970.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 ENST00000462958.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 NM_139343.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 NM_139344.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 ENST00000316724.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 ENST00000357970.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 ENST00000462958.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 NM_139343.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 NM_139344.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203394.hg.1 TC0200014160.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0