This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01119764_m1
See other CHAMP1 GE Assays ›
Gene Symbol:
CHAMP1
Gene Name:
chromosome alignment maintaining phosphoprotein 1
Gene Aliases:
C13orf8, CAMP, CHAMP, MRD40, ZNF828
Chromosome Location:
Chr.13: 114314490 - 114327328 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01119764_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHAMP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164144.1 NP_001157616.1 1 - 2 155 102
NM_032436.2 NP_115812.1 1 - 2 155 102
GenBank mRNA AK096346.1 - 1 - 2 132 102
BC065237.1 - 1 - 2 105 6162118 102
DA289328.1 - 1 - 2 124 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008181.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000437.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK096346.1,BC065237.1,NM_001164144.1,NM_032436.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHAMP1

Gene Ontology Categories:

Function(s) Process(es)

sister chromatid biorientation
protein localization to kinetochore
protein localization to microtubule
attachment of mitotic spindle microtubules to kinetochore
nucleic acid binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000463003.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000463003.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000463003.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000478022.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 ENST00000478022.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13004997.hg.1 ENST00000361283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13004997.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13004997.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13004997.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13004997.hg.1 TC13000437.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13004998.hg.1 ENST00000361283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13004998.hg.1 NM_001164144.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13004998.hg.1 NM_001164145.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13004998.hg.1 NM_032436.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13004998.hg.1 TC13000437.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay PSR1300155148.hg.1 TC1300008181.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000463003.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000463003.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000463003.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000478022.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 ENST00000478022.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay TC1300008181.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 ENST00000361283.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 NM_001164144.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 NM_001164145.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 NM_032436.3 Refseq
Affymetrix Human Clariom D Assay PSR1300155149.hg.1 TC1300008181.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 180347 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0