This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01119751_m1
See other ADGRF4 GE Assays ›
Gene Symbol:
ADGRF4
Gene Name:
adhesion G protein-coupled receptor F4
Gene Aliases:
GPR115, PGR18
Chromosome Location:
Chr.6: 47698553 - 47722021 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01119751_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRF4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153838.3 NP_722580.3 8 - 9 2292 140
GenBank mRNA AK095395.1 - 8 - 9 2204 140
DQ315370.1 - 8 - 9 2279 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600014136.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06004090.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRF4

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007749.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007749.hg.1 ENST00000327753.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007749.hg.1 ENST00000371220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007749.hg.1 NM_153838.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007749.hg.1 TC06004090.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0600198859.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600198859.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600198859.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay PSR0600198859.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600198859.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600198859.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay PSR0600198859.hg.1 TC0600014136.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000283303.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 ENST00000327753.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq
Affymetrix Human Clariom D Assay TC0600014136.hg.1 NM_153838.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0