Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007746.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007746.hg.1 |
ENST00000327753.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007746.hg.1 |
ENST00000371220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007746.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007746.hg.1 |
TC06004090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198856.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198856.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198856.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198856.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198856.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198856.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198856.hg.1 |
TC0600014136.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000283303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
ENST00000327753.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014136.hg.1 |
NM_153838.4 |
Refseq |