This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01119262_m1
See other PGF GE Assays ›
Gene Symbol:
PGF
Gene Name:
placental growth factor
Gene Aliases:
D12S1900, PGFL, PIGF, PLGF, PlGF-2, SHGC-10760
Chromosome Location:
Chr.14: 74941830 - 74955764 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01119262_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293643.1 NP_001280572.1 4 - 5 914 73
NM_002632.5 NP_002623.2 4 - 5 917 73
GenBank mRNA AL543693.3 - 4 - 5 728 73
BC001422.2 - 4 - 5 741 3139175 73
BC007255.1 - 4 - 5 722 3028008 73
BC007789.2 - 4 - 5 719 3629125 73
BT007182.1 - 4 - 5 395 73
BX248289.1 - 4 - 5 917 73
S72960.1 - 4 - 5 407 73

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Human Growth Factors Inventoried Human_Growth_Factors Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PGF

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
response to hypoxia
positive regulation of endothelial cell proliferation
sprouting angiogenesis
signal transduction
cell-cell signaling
female pregnancy
positive regulation of cell proliferation
cell differentiation
organ regeneration
cellular response to hormone stimulus
response to drug
positive regulation of angiogenesis
vascular endothelial growth factor receptor signaling pathway
positive regulation of cell division
regulation of morphogenesis of a branching structure
protein binding
growth factor activity
heparin binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000238607.10 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000405431.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000553716.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555234.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555253.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000555567.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000556939.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 ENST00000557748.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001207012.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_001293643.1 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq
Affymetrix Human Clariom D Assay TC1400009705.hg.1 NM_002632.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0