This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01118358_m1
See other LAMC1 GE Assays ›
Gene Symbol:
LAMC1
Gene Name:
laminin subunit gamma 1
Gene Aliases:
LAMB2
Chromosome Location:
Chr.1: 183023460 - 183145592 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01118358_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LAMC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002293.3 NP_002284.3 26 - 27 4732 66
GenBank mRNA BC015586.2 - 8 - 9 1089 66
DA401791.1 - 3 - 4 398 66
J03202.1 - 26 - 27 4734 66
M27654.1 - 2 - 3 299 66
X13939.1 - 4 - 5 633 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100010863.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001583.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LAMC1

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
cell adhesion
endoderm development
cell migration
extracellular matrix disassembly
extracellular matrix organization
hemidesmosome assembly
substrate adhesion-dependent cell spreading
positive regulation of epithelial cell proliferation
extracellular matrix structural constituent

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01024522.hg.1 ENST00000258341.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01024522.hg.1 NM_002293.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01024522.hg.1 TC01001583.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100179878.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100179878.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay PSR0100179878.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100179878.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay PSR0100179878.hg.1 TC0100010863.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000258341.4 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000466964.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000478064.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000479499.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000484114.2 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 ENST00000495918.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq
Affymetrix Human Clariom D Assay TC0100010863.hg.1 NM_002293.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0