This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01117609_m1
See other HERC2 GE Assays ›
Gene Symbol:
HERC2
Gene Name:
HECT and RLD domain containing E3 ubiquitin protein ligase 2
Gene Aliases:
D15F37S1, MRT38, SHEP1, jdf2, p528
Chromosome Location:
Chr.15: 28111037 - 28322173 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01117609_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HERC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004667.5 NP_004658.3 81 - 82 12670 139
XM_005268276.4 XP_005268333.1 80 - 81 12800 139
XM_006720726.3 XP_006720789.1 81 - 82 12676 139
XM_006720727.3 XP_006720790.1 79 - 80 12433 139
XM_011522131.2 XP_011520433.1 79 - 80 12457 139
XM_011522133.2 XP_011520435.1 62 - 63 9436 139
XM_017022695.1 XP_016878184.1 80 - 81 12617 139
XM_017022696.1 XP_016878185.1 80 - 81 12560 139
XM_017022697.1 XP_016878186.1 40 - 41 5999 139
XM_017022698.1 XP_016878187.1 40 - 41 5910 139
GenBank mRNA AF071172.3 - 81 - 82 12622 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500008796.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001092.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HERC2

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via nonhomologous end joining
intracellular protein transport
cellular response to DNA damage stimulus
spermatogenesis
protein ubiquitination
protein sumoylation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of GTPase activity
ubiquitin-protein transferase activity
guanyl-nucleotide exchange factor activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
SUMO binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0