This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115971_g1
See other ZMYM4 GE Assays ›
Gene Symbol:
ZMYM4
Gene Name:
zinc finger MYM-type containing 4
Gene Aliases:
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location:
Chr.1: 35268694 - 35421944 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115971_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZMYM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005095.2 NP_005086.2 6 - 7 1003 86
XM_005271328.4 XP_005271385.1 6 - 7 1282 86
XM_005271331.2 XP_005271388.1 3 - 4 248 86
XM_011542421.2 XP_011540723.1 6 - 7 929 86
XM_011542423.2 XP_011540725.1 7 - 8 1334 86
XM_011542425.2 XP_011540727.1 4 - 5 288 86
XM_017002803.1 XP_016858292.1 7 - 8 1056 86
XM_017002804.1 XP_016858293.1 7 - 8 996 86
GenBank mRNA AB007885.1 - 2 - 3 123 86
AK308320.1 - 6 - 7 1003 86
AK309705.1 - 5 - 6 663 86
BC012093.1 - 4 - 5 582 86
BC127113.1 - 7 - 8 1009 40125110 86
BC127114.1 - 3 - 4 192 40125115 86
BX647376.1 - 2 - 3 233 86
CR936741.1 - 5 - 6 867 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007765.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000456.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC127114.1,XM_005271331.2,XM_011542425.2,XM_017002804.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZMYM4

Panther Classification:

Molecular Function -

zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000364672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0