This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115965_m1
See other ZMYM4 GE Assays ›
Gene Symbol:
ZMYM4
Gene Name:
zinc finger MYM-type containing 4
Gene Aliases:
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location:
Chr.1: 35268694 - 35421944 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115965_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZMYM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005095.2 NP_005086.2 28 - 29 4382 104
XM_005271328.4 XP_005271385.1 28 - 29 4661 104
XM_005271331.2 XP_005271388.1 25 - 26 3627 104
XM_011542421.2 XP_011540723.1 28 - 29 4308 104
XM_011542423.2 XP_011540725.1 29 - 30 4713 104
XM_011542425.2 XP_011540727.1 26 - 27 3667 104
XM_017002803.1 XP_016858292.1 29 - 30 4435 104
XM_017002804.1 XP_016858293.1 29 - 30 4375 104
XM_017002805.1 XP_016858294.1 24 - 25 3873 104
GenBank mRNA AB007885.1 - 24 - 25 3502 104
AK309705.1 - 27 - 28 4042 104
BC012093.1 - 26 - 27 3961 104
BC127113.1 - 29 - 30 4388 40125110 104
BC127114.1 - 25 - 26 3571 40125115 104
BX648067.1 - 19 - 20 2676 104
CR936741.1 - 27 - 28 4246 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007765.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000456.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZMYM4

Panther Classification:

Molecular Function -

zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000364672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0