This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115961_g1
See other ZMYM4 GE Assays ›
Gene Symbol:
ZMYM4
Gene Name:
zinc finger MYM-type containing 4
Gene Aliases:
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location:
Chr.1: 35268694 - 35421944 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115961_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZMYM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005095.2 NP_005086.2 24 - 25 3781 124
XM_005271328.4 XP_005271385.1 24 - 25 4060 124
XM_005271331.2 XP_005271388.1 21 - 22 3026 124
XM_011542421.2 XP_011540723.1 24 - 25 3707 124
XM_011542423.2 XP_011540725.1 25 - 26 4112 124
XM_011542425.2 XP_011540727.1 22 - 23 3066 124
XM_017002803.1 XP_016858292.1 25 - 26 3834 124
XM_017002804.1 XP_016858293.1 25 - 26 3774 124
XM_017002805.1 XP_016858294.1 20 - 21 3272 124
GenBank mRNA AB007885.1 - 20 - 21 2901 124
AK309705.1 - 23 - 24 3441 124
BC012093.1 - 22 - 23 3360 124
BC127113.1 - 25 - 26 3787 40125110 124
BC127114.1 - 21 - 22 2970 40125115 124
BX647376.1 - 20 - 21 3012 124
BX648067.1 - 15 - 16 2075 124
CR936741.1 - 23 - 24 3645 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007765.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000456.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZMYM4

Panther Classification:

Molecular Function -

zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000364672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0