This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115947_m1
See other ZMYM4 GE Assays ›
Gene Symbol:
ZMYM4
Gene Name:
zinc finger MYM-type containing 4
Gene Aliases:
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location:
Chr.1: 35268694 - 35421944 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115947_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZMYM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005095.2 NP_005086.2 11 - 12 1911 90
XM_005271328.4 XP_005271385.1 11 - 12 2190 90
XM_005271331.2 XP_005271388.1 8 - 9 1156 90
XM_011542421.2 XP_011540723.1 11 - 12 1837 90
XM_011542423.2 XP_011540725.1 12 - 13 2242 90
XM_011542425.2 XP_011540727.1 9 - 10 1196 90
XM_017002803.1 XP_016858292.1 12 - 13 1964 90
XM_017002804.1 XP_016858293.1 12 - 13 1904 90
XM_017002805.1 XP_016858294.1 7 - 8 1402 90
GenBank mRNA AB007885.1 - 7 - 8 1031 90
AK309705.1 - 10 - 11 1571 90
BC012093.1 - 9 - 10 1490 90
BC127113.1 - 12 - 13 1917 40125110 90
BC127114.1 - 8 - 9 1100 40125115 90
BX647376.1 - 7 - 8 1141 90
BX648067.1 - 2 - 3 205 90
CR936741.1 - 10 - 11 1775 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007765.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000456.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZMYM4

Panther Classification:

Molecular Function -

zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000314607.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000364672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000441447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000457946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000470175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000482131.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 ENST00000492456.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007765.hg.1 NM_005095.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0