This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115751_m1
See other DPYD GE Assays ›
Gene Symbol:
DPYD
Gene Name:
dihydropyrimidine dehydrogenase
Gene Aliases:
DHP, DHPDHASE, DPD
Chromosome Location:
Chr.1: 97077743 - 97921059 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115751_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DPYD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000110.3 NP_000101.2 4 - 5 452 113
NM_001160301.1 NP_001153773.1 4 - 5 452 113
XM_005270562.3 XP_005270619.2 4 - 5 452 113
XM_006710397.3 XP_006710460.1 4 - 5 452 113
XM_017000507.1 XP_016855996.1 3 - 4 335 113
XM_017000508.1 XP_016855997.1 4 - 5 447 113
XM_017000509.1 XP_016855998.1 4 - 5 447 113
XM_017000510.1 XP_016855999.1 3 - 4 336 113
GenBank mRNA AB003063.1 - 4 - 5 315 113
AK291217.1 - 4 - 5 418 113
AK297515.1 - 3 - 4 279 113
BC008379.1 - 4 - 5 339 113
BC064027.1 - 4 - 5 378 113
BC108742.1 - 4 - 5 411 113
BC131777.1 - 4 - 5 368 40123875 113
BC131778.1 - 4 - 5 368 40123878 113
BT006740.1 - 4 - 5 315 113
DA721123.1 - 4 - 5 452 113
U09178.1 - 4 - 5 396 113
U20938.1 - 4 - 5 416 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015023.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002903.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017000508.1,XM_017000509.1,XM_017000510.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DPYD

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

purine nucleobase catabolic process
pyrimidine nucleobase catabolic process
thymine catabolic process
uracil catabolic process
thymidine catabolic process
beta-alanine biosynthetic process
pyrimidine nucleoside catabolic process
oxidation-reduction process
protein binding
dihydropyrimidine dehydrogenase (NADP+) activity
protein homodimerization activity
metal ion binding
flavin adenine dinucleotide binding
NADP binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000306031.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000370192.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000460019.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000460019.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000460019.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000460019.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000474241.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000474241.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000474241.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000474241.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000474241.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 ENST00000474241.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_000110.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015023.hg.1 NM_001160301.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0